QLA Bent 316 SS Cannula with Permanent Tip Compatible with Agilent and VanKel Dissolution Apparatus, 4.75"L; SS Luer Lock
Representative image only.
QLA Bent 316 SS Cannula with Permanent Tip Compatible with Agilent and VanKel Dissolution Apparatus, 4.75"L; SS Luer Lock
| Mfr: CAN475-VK| Item: 4130235
Achieve consistent sampling with QLA cannulas
₹41,600.00INR / Each
₹41,600.00INR / Each
Buy It Together
Current Item
QLA Bent 316 SS Cannula with Permanent Tip Compatible with Agilent and VanKel Dissolution Apparatus, 4.75"L; SS Luer Lock
QLA Bent 316 SS Cannula with Permanent Tip Compatible with Agilent and VanKel Dissolution Apparatus, 4.75"L; SS Luer Lock
4130235
₹41,600.00INR / Each
+ =
Total Price 58900.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Matching cannula to stopper ensures greater sampling position consistency
  • 4.75"L with permanent tip
More About this Item
For dissolution tests, taking reliable samples is critical. QLA probes, cannulas and stoppers ensure consistent sampling position within the mixing vessel, which means more accurate testing.
Parts are made of 316 stainless steel that will not absorb or interfere with active ingredients.
.
Specifications
  • Material316 stainless steel
  • Length (")4 3/4
  • DescriptionBent 316 SS Cannula with Permanent Tip Compatible with Agilent and VanKel Dissolution Apparatus, 4.75"L; SS Luer Lock