QLA Bent 316 SS Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, Thru Head Mount, 15"L, 1/8" OD; SS Luer Lock
Representative image only.
QLA Bent 316 SS Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, Thru Head Mount, 15"L, 1/8" OD; SS Luer Lock
| Mfr: CA8015-VK| Item: 4130202
Achieve consistent sampling with QLA cannulas
₹34,000.00INR / Each
₹34,000.00INR / Each
Buy It Together
Current Item
QLA Bent 316 SS Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, Thru Head Mount, 15"L, 1/8" OD; SS Luer Lock
QLA Bent 316 SS Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, Thru Head Mount, 15"L, 1/8" OD; SS Luer Lock
4130202
₹34,000.00INR / Each
+ =
Total Price 51300.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Matching cannula to stopper ensures greater sampling position consistency
  • 15" long
  • Thru mount
  • 316 SS construction
More About this Item
For dissolution tests, taking reliable samples is critical. QLA probes, cannulas and stoppers ensure consistent sampling position within the mixing vessel, which means more accurate testing.
Parts are made of 316 stainless steel that will not absorb or interfere with active ingredients.
.
Specifications
  • Material316 stainless steel
  • Outside Diameter (OD) (mm)3.2
  • Length (")15
  • DescriptionBent 316 SS Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, Thru Head Mount, 15"L, 1/8" OD; SS Luer Lock