QLA Bent 316 SS Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, for 500 mL vessel, 7.75"L, 1/8" OD; SS Luer Lock
Representative image only.
QLA Bent 316 SS Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, for 500 mL vessel, 7.75"L, 1/8" OD; SS Luer Lock
| Mfr: CA8775-VK| Item: 4130207
Achieve consistent sampling with QLA cannulas
₹29,500.00INR / Each
₹29,500.00INR / Each
Buy It Together
Current Item
QLA Bent 316 SS Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, for 500 mL vessel, 7.75"L, 1/8" OD; SS Luer Lock
QLA Bent 316 SS Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, for 500 mL vessel, 7.75"L, 1/8" OD; SS Luer Lock
4130207
₹29,500.00INR / Each
+ =
Total Price 46800.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Matching cannula to stopper ensures greater sampling position consistency
  • 7.75"L for 500 mL vessel
  • 316 SS construction
  • 1/8" OD
More About this Item
For dissolution tests, taking reliable samples is critical. QLA probes, cannulas and stoppers ensure consistent sampling position within the mixing vessel, which means more accurate testing.
Parts are made of 316 stainless steel that will not absorb or interfere with active ingredients.
.
Specifications
  • Material316 stainless steel
  • Outside Diameter (OD) (mm)3.2
  • Length (")7 3/4
  • DescriptionBent 316 SS Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, for 500 mL vessel, 7.75"L, 1/8" OD; SS Luer Lock