QLA Bent 316 SS Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 7.75"L; SS Luer Lock and Perm SS Tip
Representative image only.
QLA Bent 316 SS Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 7.75"L; SS Luer Lock and Perm SS Tip
| Mfr: CAN775-VK| Item: 4130252
Achieve consistent sampling with QLA cannulas
₹42,100.00INR / Each
₹42,100.00INR / Each
Buy It Together
Current Item
QLA Bent 316 SS Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 7.75"L; SS Luer Lock and Perm SS Tip
QLA Bent 316 SS Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 7.75"L; SS Luer Lock and Perm SS Tip
4130252
₹42,100.00INR / Each
+ =
Total Price 59400.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Matching cannula to stopper ensures greater sampling position consistency
  • 7.75"L with permanent stainless steel tip
More About this Item
For dissolution tests, taking reliable samples is critical. QLA probes, cannulas and stoppers ensure consistent sampling position within the mixing vessel, which means more accurate testing.
Parts are made of 316 stainless steel that will not absorb or interfere with active ingredients.
.
Specifications
  • Material316 stainless steel
  • Length (")7 3/4
  • DescriptionBent 316 SS Cannula Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 7.75"L; SS Luer Lock and Perm SS Tip