QLA Bent 316 SS Cannula Compatible with Agilent and VanKel Dissolution Apparatus, for 500 mL vessel, 7.75"L, 0.082" OD; SS Luer Lock
Representative image only.
QLA Bent 316 SS Cannula Compatible with Agilent and VanKel Dissolution Apparatus, for 500 mL vessel, 7.75"L, 0.082" OD; SS Luer Lock
| Mfr: CAN775-01| Item: 4130245
Achieve consistent sampling with QLA cannulas
₹44,500.00INR / Each
₹44,500.00INR / Each
Buy It Together
Current Item
QLA Bent 316 SS Cannula Compatible with Agilent and VanKel Dissolution Apparatus, for 500 mL vessel, 7.75"L, 0.082" OD; SS Luer Lock
QLA Bent 316 SS Cannula Compatible with Agilent and VanKel Dissolution Apparatus, for 500 mL vessel, 7.75"L, 0.082" OD; SS Luer Lock
4130245
₹44,500.00INR / Each
+ =
Total Price 61800.00 ADD 2 ITEMS TO CART
Product Description
Includes
Tygon® tubing.
Key Features
  • Matching cannula to stopper ensures greater sampling position consistency
  • 7.75"L for 900 mL vessels
  • SS luer lock
  • 0.082" OD
More About this Item
For dissolution tests, taking reliable samples is critical. QLA probes, cannulas and stoppers ensure consistent sampling position within the mixing vessel, which means more accurate testing.
Parts are made of 316 stainless steel that will not absorb or interfere with active ingredients.
.
Specifications
  • Material316 stainless steel
  • Outside Diameter (OD) (mm)2.1
  • Length (")7 3/4
  • DescriptionBent 316 SS Cannula Compatible with Agilent and VanKel Dissolution Apparatus, for 500 mL vessel, 7.75"L, 0.082" OD; SS Luer Lock