QLA Adjustable Cannula Stopper, 0.082" OD; with Nut and Ferrule
Representative image only.
QLA Adjustable Cannula Stopper, 0.082" OD; with Nut and Ferrule
| Mfr: CANSTP-DVN| Item: 4130278
QLA accessories help provide more precise and consistent dissolution results
₹12,400.00INR / Each
₹12,400.00INR / Each
Product Description
Key Features
  • 0.082" OD
  • Comes with nut and ferrule
More About this Item
Use stopper with cannula to ensure the sample is always taken from the same depth.
.
Specifications
  • Compatible WithAny brand
  • DescriptionAdjustable Cannula Stopper, 0.082" OD; with Nut and Ferrule