QLA Bent 316 SS Cannula Compatible with Agilent and VanKel Dissolution Apparatus, for 500 mL vessel, 7.75"L, 0.082" OD; PVDF Luer Lock
Representative image only.
QLA Bent 316 SS Cannula Compatible with Agilent and VanKel Dissolution Apparatus, for 500 mL vessel, 7.75"L, 0.082" OD; PVDF Luer Lock
| Mfr: CAN775-01L| Item: 4130246
Achieve consistent sampling with QLA cannulas
₹18,500.00INR / Each
₹18,500.00INR / Each
Buy It Together
Current Item
QLA Bent 316 SS Cannula Compatible with Agilent and VanKel Dissolution Apparatus, for 500 mL vessel, 7.75"L, 0.082" OD; PVDF Luer Lock
QLA Bent 316 SS Cannula Compatible with Agilent and VanKel Dissolution Apparatus, for 500 mL vessel, 7.75"L, 0.082" OD; PVDF Luer Lock
4130246
₹18,500.00INR / Each
+ =
Total Price 35800.00 ADD 2 ITEMS TO CART
Product Description
Includes
Tygon® tubing.
Key Features
  • Matching cannula to stopper ensures greater sampling position consistency
  • 7.75"L for 900 mL vessels
  • PVDF luer lock
  • 0.082" OD
More About this Item
For dissolution tests, taking reliable samples is critical. QLA probes, cannulas and stoppers ensure consistent sampling position within the mixing vessel, which means more accurate testing.
316 stainless steel cannula with PVDF luer lock offers cost savings.
.
Specifications
  • Material316 SS cannula/PVDF luer lock
  • Outside Diameter (OD) (mm)2.1
  • Length (")7 3/4
  • DescriptionBent 316 SS Cannula Compatible with Agilent and VanKel Dissolution Apparatus, for 500 mL vessel, 7.75"L, 0.082" OD; PVDF Luer Lock