QLA Auto-Sampling Cannula Compatible with Agilent and VanKel Dissolution Apparatus, 6.5"L; for 500mL Sampling
Representative image only.
QLA Auto-Sampling Cannula Compatible with Agilent and VanKel Dissolution Apparatus, 6.5"L; for 500mL Sampling
| Mfr: CAN500-60| Item: 4130236
Achieve consistent sampling with QLA cannulas
₹16,500.00INR / Each
₹16,500.00INR / Each
Buy It Together
Current Item
QLA Auto-Sampling Cannula Compatible with Agilent and VanKel Dissolution Apparatus, 6.5"L; for 500mL Sampling
QLA Auto-Sampling Cannula Compatible with Agilent and VanKel Dissolution Apparatus, 6.5"L; for 500mL Sampling
4130236
₹16,500.00INR / Each
+ =
Total Price 33800.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Matching cannula to stopper ensures greater sampling position consistency
  • 6.5"L for auto sampling station
More About this Item
For dissolution tests, taking reliable samples is critical. QLA probes, cannulas and stoppers ensure consistent sampling position within the mixing vessel, which means more accurate testing.
Parts are made of 316 stainless steel that will not absorb or interfere with active ingredients.
.
Specifications
  • Material316 stainless steel
  • Length (")6 1/2
  • DescriptionAuto-Sampling Cannula Compatible with Agilent and VanKel Dissolution Apparatus, 6.5"L; for 500mL Sampling