PRO Scientific Multi-Gen 7XL Motor Unit Adapter
Representative image only.
PRO Scientific Multi-Gen 7XL Motor Unit Adapter
PRO Scientific | Mfr: 07-07200| Item: 0475169
Expand and upgrade your homogenizing capabilities
₹131,000.00INR / Each
₹131,000.00INR / Each
Buy It Together
Current Item
PRO Scientific Multi-Gen 7XL  Motor Unit Adapter
PRO Scientific Multi-Gen 7XL Motor Unit Adapter
0475169
₹131,000.00INR / Each
+ + =
Total Price 709000.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 2 year warranty
More About this Item
Parts and accessories compatible with PRO Scientific homogenizers.
Multi-Gen 7XL Generator Probe System combines an adapter (04751-69) and a pack of twelve 316 stainless steel and PTFE generators (04751-70) to replace disposable plastic generators. Adapter is compatible with most PRO Scientific homogenizer models including Bio-Gen PRO200, PRO250, PRO25D, SB-1, and PRO400DEL. The generator probes feature an open slotted tooth design suitable for processing even tough tissue samples up to 15 mL, and can be sterilized by autoclave or flame. Unlimited reusability make this system an economical choice when cross-contamination is a serious concern.
Two year warranty
Specifications
  • MaterialStainless Steel
  • Compatible WithPRO Scientific Homogenizers
  • Diameter (mm)7
  • DescriptionMotor Unit Adapter