PRO Scientific Bio-Gen PRO200 Homogenizer
Small and powerful
  • Compact handheld design processes samples in tubes and small beakers
  • Variable-speed motor provides enough torque to homogenize even tough tissues
This handheld homogenizer is a versatile tool, combining an easy-to-hold housing with a powerful 144-watt motor. For continuous processing, there is a separate on/off switch and the homogenizer can be post-mounted using the optional stand assembly (order separately).
Choose a generator probe based on the sample volume and dispersion type (order separately). The standard quick-connect generators are precision crafted from 316 stainless steel and PTFE. The flat-bottom probe is for small volumes and the saw-tooth creates enough shear to handle larger volumes and tough tissues. The Multi-Gen system offers low-cost repeat homogenization without the risk of cross-contamination. Clean and sterilize the 316 stainless steel and PTFE Multi-Gen probes.
223,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
PRO Scientific Bio-Gen PRO200 Homogenizer, Handheld or Post Mounted; 120 V WW-04751-20 Mfr # 01-01200
COMPARE
120
₹223,000.00INR / Each
PRO Scientific Bio-Gen PRO200 Homogenizer, Handheld or Post Mounted; 220 V WW-04751-25 Mfr # 01-02200
COMPARE
220
₹223,000.00INR / Each