PendoTech Single-Use Temperature Sensor, Non-Sterile, Polysulfone, 1/8" Hose Barb
Representative image only.
PendoTech Single-Use Temperature Sensor, Non-Sterile, Polysulfone, 1/8" Hose Barb
PendoTech | Mfr: TEMPS-N-012| Item: 5990023
Accurate, cost-effective, and single use for elimination of cross-contamination
₹15,300.00INR / Each
₹15,300.00INR / Each
Buy It Together
Current Item
PendoTech Single-Use Temperature Sensor, Non-Sterile, Polysulfone, 1/8" Hose Barb
PendoTech Single-Use Temperature Sensor, Non-Sterile, Polysulfone, 1/8" Hose Barb
5990023
₹15,300.00INR / Each
+ + + +
PendoTech PressureMAT® DPG Handheld Pressure Monitor
5990009
₹204,000.00INR / Each
+ =
Total Price 986800.00 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • No calibration required—each sensor is pretested for accuracy
  • High sensitivity with better than ±0.2°C accuracy in the 0 to 70°C range
  • Biocompatible materials meeting USP Class VI standards
  • Gamma and x-ray irradiation compatible for sterilization
  • 2 year warranty
More About this Item
PendoTECH® Single-Use Temperature Sensors™ deliver precise and reliable in-line temperature measurements for bioprocess applications such as filtration, chromatography, filling operations, and general process monitoring. Connect sensors to a transmitter via a reusable cable; transmitter provides a 4 to 20 mA output signal for an easy connection to PressureMAT® Plus for real-time monitoring (interface cable required).
Sensors feature hose barb fittings to prevent pressure drops and dead legs, and require no calibration, ensuring easy setup and dependable performance. Designed to minimize contamination risks while maintaining high accuracy (±0.2°C or better), they are available in multiple sizes to integrate smoothly into existing workflows. Manufactured in an ISO 9001-certified facility, these sensors provide a cost-effective alternative to traditional temperature monitoring devices while supporting single-use bioprocessing needs.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Two year warranty
Specifications
  • Accuracy±0.2°C
  • Min Temperature (° F)32
  • Max Temperature (° F)158
  • Min Temperature (° C)0
  • Max Temperature (DegreeC)70
  • Wetted MaterialsPolysulfone
  • Process Connection1/8" Hose Barb
  • Electrical Connections10-ft cable with 1/4" headphone plug
  • Media CompatibilityAll materials in contact with fluid path meet USP Class VI
  • SterileNo
  • DescriptionSingle-Use Temperature Sensor, Non-Sterile, Polysulfone, 1/8" Hose Barb
  • Warranty2 years