PendoTECH® PressureMAT® Plus Monitors
Advanced monitoring, alarm, and transmitter system for bioprocess control
  • Compatible with PendoTECH® Single-Use Pressure Sensors™ and Flow Meters
  • 4-20 mA analog input function for external transmitter integration
  • Real-time monitoring of pressure, flow rate, and total flow volume
  • User-configurable min/max alarm set points with alarm output signal
  • Data output to PC for integration with PendoTECH® Data Acquisition Software
The PendoTECH® PressureMAT® Plus is a multi-functional monitoring and control system designed for critical bioprocess applications such as filtration and chromatography. This advanced unit allows real-time monitoring of pressure, flow rate, and total flow volume with seamless integration of external 4-20 mA transmitters for additional process parameters like temperature, UV absorbance, conductivity, and turbidity. Featuring user-configurable alarms, data logging, and RS-232 output, it enables precise process control while ensuring compliance with industry standards. The IP66/NEMA 4X front panel and panel-mount options enhance durability for demanding environments.
580,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Output Process Connection
Availability Pricing
PendoTech PressureMAT® Plus Monitor for 2 Single-Use Pressure Sensors, psi 5990005 Mfr # PMAT2P
COMPARE
4 analog 2 pressure sensor inputs, 1 flow meter input, 4-20 mA input
₹580,000.00INR / Each
PendoTech PressureMAT® Plus Monitor for 3 Single-Use Pressure Sensors, psi 5990007 Mfr # PMAT3P
COMPARE
4 analog 3 pressure sensor inputs, 1 flow meter input
₹580,000.00INR / Each