PendoTech Cable, Single-Use Temperature Sensor to Transmitter, 1/4" Phone Jack, 3 m
Representative image only.
PendoTech Cable, Single-Use Temperature Sensor to Transmitter, 1/4" Phone Jack, 3 m
PendoTech | Mfr: PDKT-650-TEMPB| Item: 4852482
Cables for seamless integration of Single-Use Temperature Sensors™ to transmitters
₹21,400.00INR / Each
₹21,400.00INR / Each
Buy It Together
Current Item
PendoTech Cable, Single-Use Temperature Sensor to Transmitter, 1/4" Phone Jack, 3 m
PendoTech Cable, Single-Use Temperature Sensor to Transmitter, 1/4" Phone Jack, 3 m
4852482
₹21,400.00INR / Each
+ =
Total Price 33600.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 3-m (10-ft) reusable temperature sensor cable with 1/4" phone jack for quick connection to the transmitter
  • Use interface cable to transmit 4 to 20 mA signal to PressureMAT® Plus monitor
  • Connect temperature sensor to the transmitter
  • 1 year warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Compatible WithSingle-Use Temperature Sensor™ and benchtop transmitter
  • Connections1/4" Phone jack
  • DescriptionCable, Single-Use Temperature Sensor to Transmitter, 1/4" Phone Jack, 3 m