PendoTech Cable Adaptor with D15 for Single Use Pressure Sensor for PMAT, 12' (4 m)
Representative image only.
PendoTech Cable Adaptor with D15 for Single Use Pressure Sensor for PMAT, 12' (4 m)
PendoTech | Mfr: PMAT-650-298| Item: 2121410
Seamless integration for Single-Use Pressure Sensors™
₹24,700.00INR / Each
₹24,700.00INR / Each
Buy It Together
Current Item
PendoTech Cable Adaptor with D15 for Single Use Pressure Sensor for PMAT, 12' (4 m)
PendoTech Cable Adaptor with D15 for Single Use Pressure Sensor for PMAT, 12' (4 m)
2121410
₹24,700.00INR / Each
+ +
PendoTech PressureMAT® DPG Handheld Pressure Monitor
5990009
₹204,000.00INR / Each
+ + =
Total Price 815300.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Universal compatibility with PendoTECH® Single-Use Pressure Sensors™
  • Polycarbonate straight connectors with luer ports for secure, adaptable tubing connections
  • 12-ft cable adapter for connection to PendoTECH® sensors or other devices
More About this Item
Optimize your PendoTECH® Single-Use Pressure Sensor™ setup with precision-engineered fittings and accessories. Our polycarbonate straight connectors - available in 1/4”, 3/8”, and 1/2” sizes - feature integrated luer ports for seamless tubing adaptation and fluid transfer in bioprocess applications. Additionally, the 12-ft cable adapter enables direct sensor connectivity to third-party transmitters, control systems, and custom circuits. Engineered with high-quality shielding, insulation, and flexible design, this adapter ensures reliable data transmission in critical processing environments.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Compatible WithPendoTech PressureMAT® Sensor Monitor
  • Length (InchShort)144
  • Length (cm)365.76
  • DescriptionCable Adaptor with D15 for Single Use Pressure Sensor for PMAT, 12' (4 m)