PendoTECH® PressureMAT® Sensor Monitor
Precision pressure monitoring for critical bioprocess applications
  • Designed for PendoTECH Single-Use Pressure Sensors™ for seamless integration
  • Portable, lightweight design for flexible use in labs and pilot plants
  • User-configurable min/max alarm set-points with relay output for process control
  • Data output via RS-232 for integration with PCs and control systems
The PendoTECH® PressureMAT® Sensor Monitor is a versatile, high-precision unit designed for real-time monitoring of pressure in filtration, chromatography, bioreactor monitoring, perfusion, and fill finish operations. With support for single-use pressure sensors, it provides continuous monitoring, one-touch zero tare functionality, and high-resolution readings. The system includes user-configurable alarms and relay output for automated control of pumps and valves. Featuring IP66/NEMA 4X protection and a data acquisition interface, the PressureMAT® ensures reliable and accurate pressure measurement for biopharmaceutical applications.
294,000.00 - ₹580,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Output Process Connection
Availability Pricing
PendoTech PressureMAT® Sensor Monitor with 3 Pressure Sensor Inputs and 4 Outputs, psi 5990006 Mfr # PMAT3
COMPARE
1 relay and 3 analog 3 pressure sensor inputs
₹561,000.00INR / Each
PendoTech PressureMAT® Sensor Monitor with 4 Pressure Sensor Inputs and 4 Outputs, psi 5990008 Mfr # PMAT4A
COMPARE
4 analog 4 pressure sensor inputs
₹580,000.00INR / Each
PendoTech PressureMAT® Sensor Monitor with 1 Pressure Sensor Input and 2 Outputs, psi 5990010 Mfr # PMAT-S
COMPARE
1 relay and 1 analog 1 pressure sensor input
₹294,000.00INR / Each