PendoTech 3/8" x 3/8" Polycarbonate Straight Connector with Luer Port
Representative image only.
PendoTech 3/8" x 3/8" Polycarbonate Straight Connector with Luer Port
PendoTech | Mfr: PDKT-104-03| Item: 5990002
Seamless integration for Single-Use Pressure Sensors™
₹970.00INR / Each
₹970.00INR / Each
Buy It Together
Current Item
PendoTech 3/8" x 3/8" polycarbonate straight connector with luer port
PendoTech 3/8" x 3/8" polycarbonate straight connector with luer port
5990002
₹970.00INR / Each
+ +
PendoTech PressureMAT® DPG Handheld Pressure Monitor
5990009
₹204,000.00INR / Each
+ + =
Total Price 810570.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Universal compatibility with PendoTECH® Single-Use Pressure Sensors™
  • Polycarbonate straight connectors with luer ports for secure, adaptable tubing connections
  • 12-ft cable adapter for connection to PendoTECH® sensors or other devices
  • 2 year warranty
More About this Item
Optimize your PendoTECH® Single-Use Pressure Sensor™ setup with precision-engineered fittings and accessories. Our polycarbonate straight connectors - available in 1/4”, 3/8”, and 1/2” sizes - feature integrated luer ports for seamless tubing adaptation and fluid transfer in bioprocess applications. Additionally, the 12-ft cable adapter enables direct sensor connectivity to third-party transmitters, control systems, and custom circuits. Engineered with high-quality shielding, insulation, and flexible design, this adapter ensures reliable data transmission in critical processing environments.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Two year warranty
Specifications
  • MaterialPolycarbonate
  • Compatible WithPendoTECH® Single-Use Pressor Sensors™
  • Description3/8" x 3/8" Polycarbonate Straight Connector with Luer Port
  • Warranty2 years