Omni International 19-620 2.38 mm Stainless Steel bead kit for Bead Ruptor 12
Representative image only.
Omni International 19-620 2.38 mm Stainless Steel bead kit for Bead Ruptor 12
Omni International | Mfr: 19-620| Item: 0472819
Prevent cross-contamination with disposable tubes
₹54,000.00INR / Pkg of 50
₹54,000.00INR / Pkg of 50
Buy It Together
Current Item
Omni International 19-620 2.38 mm Stainless Steel bead kit for Bead Ruptor 12
Omni International 19-620 2.38 mm Stainless Steel bead kit for Bead Ruptor 12
0472819
₹54,000.00INR / Pkg of 50
+ +
Omni International 19-648 2 mL empty sample tubes with caps, 500/pack
0472841
₹131,000.00INR / Pkg of 500
=
Total Price 185000.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ideal for use with Omni Bead Ruptor 12 homogenizer
More About this Item
Five different bead types allow processing of different samples in your Omni Bead Ruptor 12 homogenizer, 04728-01 (sold separately). Ceramic beads are ideal for soft animal, plant or human tissues. Metal beads are ideal for hard tissue types. Glass beads are ideal for processing microorganisms. Garnet beads are ideal for soil. Carbide beads are ideal for microbial and soil RNA processing.
Specifications
  • Description2.38 mm Stainless Steel bead kit for Bead Ruptor 12