Myron L RE-10 Range extender for meters
Representative image only.
Myron L RE-10 Range extender for meters
Myron L | Mfr: RE-10| Item: 0148950
Optimize water analysis
₹9,600.00INR / Each
₹9,600.00INR / Each
Buy It Together
Current Item
Myron L RE-10 Range extender for meters
Myron L RE-10 Range extender for meters
0148950
₹9,600.00INR / Each
+
Myron L 300 QCP (NIST) conductivity calibration standard, 445 µS
0148993
₹8,400.00INR / Quart of 1
+ + + + =
Total Price 988000.00 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose from a variety of accessories
  • Extends the range of your instrument by a factor of 10
  • 1 year warranty
More About this Item
Keep your Myron L meter and system running smoothly and accurately with these parts and accessories.
The RE-10 range extender is ideal for testing high conductivity/ppm solutions beyond the normal range of your meter. To use, simply fill the cell cup with your sample and push the range extender into the cup, seating the o-ring seal. Use and read your DS meter as normal, multiplying the reading by 10. Once done, remove the range extender from the sample cup and rinse the cup and extender with distilled or deionized water to eliminate salt buildup.
Do not use the range extender with model 512T10.
1 Year Warranty
Specifications
  • DescriptionRange extender for meters