Myron L CS51 Conductivity cell, K = 1.0, built-in thermistor ATC, 3/4" NPT(M) thread, 10-ft cable
Representative image only.
Myron L CS51 Conductivity cell, K = 1.0, built-in thermistor ATC, 3/4" NPT(M) thread, 10-ft cable
Myron L | Mfr: CS51| Item: 0597050
Optimize water analysis
₹67,500.00INR / Each
₹67,500.00INR / Each
Buy It Together
Current Item
Myron L CS51 Conductivity cell, K = 1.0, built-in thermistor ATC, 3/4" NPT(M) thread, 10-ft cable
Myron L CS51 Conductivity cell, K = 1.0, built-in thermistor ATC, 3/4" NPT(M) thread, 10-ft cable
0597050
₹67,500.00INR / Each
+ + + + +
Myron L 300 QCP (NIST) conductivity calibration standard, 445 µS
0148993
₹8,400.00INR / Quart of 1
=
Total Price 1045900.00 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose from a variety of accessories
  • 1 year warranty
More About this Item
Keep your Myron L meter and system running smoothly and accurately with these parts and accessories.
1 Year Warranty
Specifications
  • MaterialPolypropylene/316 SS
  • Compatible WithMyron L conductivity/TDS controllers 19010-16, 19010-18 & 19010-20
  • Length (InchShort)1 1/4
  • Diameter (InchShort)1/2
  • DescriptionConductivity cell, K = 1.0, built-in thermistor ATC, 3/4" NPT(M) thread, 10-ft cable