Myron L CS10-T Resistivity Cell, titanium, K = 0.05, 3/4" NPT(M) thread, thermistor ATC, 10ft cable
Representative image only.
Myron L CS10-T Resistivity Cell, titanium, K = 0.05, 3/4" NPT(M) thread, thermistor ATC, 10ft cable
Myron L | Mfr: CS10-T| Item: 0597072
Optimize water analysis
₹109,000.00INR / Each
₹109,000.00INR / Each
Buy It Together
Current Item
Myron L CS10-T Resistivity Cell, titanium, K = 0.05, 3/4" NPT(M) thread, thermistor ATC, 10ft cable
Myron L CS10-T Resistivity Cell, titanium, K = 0.05, 3/4" NPT(M) thread, thermistor ATC, 10ft cable
0597072
₹109,000.00INR / Each
+
Myron L 300 QCP (NIST) conductivity calibration standard, 445 µS
0148993
₹8,400.00INR / Quart of 1
+ + + + =
Total Price 1087400.00 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose from a variety of accessories
  • 1 year warranty
More About this Item
Keep your Myron L meter and system running smoothly and accurately with these parts and accessories.
1 Year Warranty
Specifications
  • MaterialPolypropylene/Titanium
  • Compatible WithMyron L resistivity controller, 19010-03
  • Length (InchShort)1 1/4
  • Diameter (InchShort)1/2
  • DescriptionResistivity Cell, titanium, K = 0.05, 3/4" NPT(M) thread, thermistor ATC, 10ft cable