Myron L 3SE Three-position switchbox (Allows one controller to read 3 cells)
Representative image only.
Myron L 3SE Three-position switchbox (Allows one controller to read 3 cells)
Myron L | Mfr: 3SE| Item: 0597082
Optimize water analysis
₹86,000.00INR / Each
₹86,000.00INR / Each
Buy It Together
Current Item
Myron L 3SE Three-position switchbox (Allows one controller to read 3 cells)
Myron L 3SE Three-position switchbox (Allows one controller to read 3 cells)
0597082
₹86,000.00INR / Each
+
Myron L 300 QCP (NIST) conductivity calibration standard, 445 µS
0148993
₹8,400.00INR / Quart of 1
+ + + + =
Total Price 1064400.00 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose from a variety of accessories
  • 1 year warranty
More About this Item
Keep your Myron L meter and system running smoothly and accurately with these parts and accessories.
Use the three-position switchbox to monitor up to three cells from the same controller. Controller relays only respond to the selected channel.
1 Year Warranty
Specifications
  • Compatible WithMyron L resistivity, conductivity/TDS controllers 19010-02, 19010-16, 19010-18 & 19010-20
  • DescriptionThree-position switchbox (Allows one controller to read 3 cells)