MTC Bio LinkLabel™ Bluetooth Enabled Laboratory Label Printer with US, UK, AU and EU plugs
Representative image only.
MTC Bio LinkLabel™ Bluetooth Enabled Laboratory Label Printer with US, UK, AU and EU plugs
MTC Bio | Mfr: L3000| Item: 0653050
Customize labels and send to printer all from your phone
₹101,000.00INR / Each
₹101,000.00INR / Each
Buy It Together
Current Item
MTC Bio LinkLabel™  Bluetooth Enabled Laboratory Label Printer with US, UK, AU and EU plugs
MTC Bio LinkLabel™ Bluetooth Enabled Laboratory Label Printer with US, UK, AU and EU plugs
0653050
₹101,000.00INR / Each
+ =
Total Price 109600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
One 26' cassette of 18mm lab tape, white w/ black print and US, UK, AU, and EU power plugs.
Key Features
  • Compatible with Bluetooth devices
  • App allows for label customization
  • Prints barcodes, QR codes, serial numbers, and batches
  • 2 year warranty
More About this Item
This compact lab label printer with Bluetooth allows you the freedom to customize and create specialize labels from your phone or other compatible devices. Hundreds of preloaded scientific symbols are available. MTC’s free LabelPro App allows you to easily change text, font, positioning, size, and label length. The label printer can print batches, serial numbers, QR codes, and barcodes.
Choose from 13 label cassettes (sold separately) available in a variety of tape colors, including transparent. Three label heights are available for different sized vessels. The 6 mm tapes are great for tissue culture plates, 12 mm tapes are ideal from microtubes, and the extra tall 24 mm tapes are ideal for large tubes and flasks. Special "ultra-low" label tapes are also available for storage in -80 °C environments, or for liquid nitrogen applications.
Conformite EuropeenneTwo year warranty
Specifications
  • ColorBlue/Black
  • Length (InchShort)5 1/4
  • Height (in)6.75
  • Width (InchShort)2 3/4
  • Power (VAC)115 / 230
  • DescriptionBluetooth Enabled Laboratory Label Printer
  • Warranty2 years