MTC Bio 20' Cassette of 24mm Ultra Low Temperature Tape, White w/ Black Print
Representative image only.
MTC Bio 20' Cassette of 24mm Ultra Low Temperature Tape, White w/ Black Print
MTC Bio | Mfr: L9010-24ULT| Item: 0653062
Ideal label identification for a variety of laboratory applications
₹14,200.00INR / Each
₹14,200.00INR / Each
Buy It Together
Current Item
MTC Bio   20' Cassette of 24mm Ultra Low Temperature Tape, White w/ Black Print
MTC Bio 20' Cassette of 24mm Ultra Low Temperature Tape, White w/ Black Print
0653062
₹14,200.00INR / Each
+ + =
Total Price 175700.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Can be autoclaved, boiled, or frozen
  • Manufacturer defect warranty
More About this Item
These label printer cassette tapes are ideal for label identification for nearly any laboratory application. Compatible with MTC Bio handheld and Bluetooth® enable laboratory label printers, these tapes can be autoclaved, boiled or frozen to temperatures as low as -50 °C.
Choose from 13 label cassettes tapes available in a variety of colors, including transparent. Three label heights are available for different sized vessels. The 6 mm tapes are great for tissue culture plates, 12 mm tapes are ideal from microtubes, and the extra tall 24 mm tapes are ideal for large tubes and flasks.
Special "ultra-low" label tapes (models 06530-57 & 06530-62) are available for storage in -80 °C environments, or for liquid nitrogen applications.
Specifications
  • ColorWhite with black print
  • Compatible WithMTC Bio handheld and Bluetooth® enable laboratory label printers
  • Length (InchShort)240
  • Width (mm)24
  • Description20' Cassette of 24mm Ultra Low Temperature Tape, White w/ Black Print
  • WarrantyManufacturer defect