Microscope Camera with SD Card, C-Mount, Auto-Focus, 2 MP, HDMI and Wi-Fi
Representative image only.
Microscope Camera with SD Card, C-Mount, Auto-Focus, 2 MP, HDMI and Wi-Fi
| Item: 7890518
Capture images and stream video directly from your microscope
₹235,000.00INR / Each
₹235,000.00INR / Each
Buy It Together
Current Item
Microscope Camera with SD Card, C-Mount, Auto-Focus, 2 MP, HDMI and Wi-Fi
Microscope Camera with SD Card, C-Mount, Auto-Focus, 2 MP, HDMI and Wi-Fi
7890518
₹235,000.00INR / Each
+ + + +
Microscope Camera Adapter for Nikon SLR/DSLR, 2X
7890500
₹32,800.00INR / Each
=
Total Price 1195800.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Includes
digital camera, USB mouse, USB wi-fi dongle, HDMI cable, SD card, power adapter, and software CD.
Key Features
  • Documentation and analysis for a deeper view of your slide
  • Easy to install and ready for immediate use
  • Capture, annotate, measure or evaluate still images and video
  • Auto-focus digital camera features frame rate of 60 fps at 1920 x 1080
More About this Item
AmScope™ microscope cameras are designed for ease of use and the ability to output images and video. These cameras can produce still image and video capture, still image measurement, and allow you to make notes, shapes, and watermarks with ease.
Cameras are available in a wide range of resolution and connectivity options for applications ranging from education, conservation, pharmaceutical, or other demanding fields. Microscopes sold separately.
Designed using the C-mount standard, the camera with auto-focus capabilities offers 1080p live-viewing over HDMI, and can record photos and video directly to Secure Digital media. The addition of auto-focus limits the need to make focus-adjustments when using the camera as the primary means of observation. The included USB mouse allows you to control the camera through its easy-to-use interface. As an alternative solution, connect to the camera over Wi-Fi for remote software control, with 25 fps streaming. The camera’s C-mount interface makes it compatible with a wide range of reduction lenses and photo-ports. Note: because of the auto-focus mechanism, C-mount lens threads should not exceed 5mm.
1 Year Warranty
Specifications
  • Resolution2 MP, 1920 x 1080
  • ObjectivesNone
  • Image Sensor1/1.9" Aptina AR1820 color CMOS
  • OutputHDMI, Wi-Fi
  • System RequirementsWindows (32/64 bit) XP/Vista/7/8/10, Mac OS 10.8+, Linux kernel 3.13+
  • Data TransferHDMI and Wi-Fi
  • Power12 VDC
  • DescriptionMicroscope Camera with SD Card, C-Mount, Auto-Focus, 2 MP, HDMI and Wi-Fi