AmScope™ Microscope Camera Adapters
Easily connect your microscope to a camera
  • Magnification power of 2X
  • Metal construction
  • Works on monocular (eye tube), binocular (eye tube) or trinocular photo/video port
AmScope microscope camera adapters are designed to connect a Nikon or Canon SLR/DSLR camera to your microscope for image capture. With two adapters included, it offers three microscope mounting sizes, 23.2 mm, 30 mm, and 30.5 mm, fitting most compound and stereo microscopes. Camera and microscopes sold separately.
32,800.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Magnification Size
Availability Pricing
COMPARE
2X Microscope Mounting Size: 23.2, 30, 30.5 mm
₹32,800.00INR / Each
COMPARE
2X Microscope Mounting Size: 23.2, 30, 30.5 mm
₹32,800.00INR / Each