Microscope Camera with Reduction Lens, C-Mount, 10 MP, USB 2.0
Representative image only.
Microscope Camera with Reduction Lens, C-Mount, 10 MP, USB 2.0
| Item: 7890514
Capture images and stream video directly from your microscope
₹93,100.00INR / Each
₹93,100.00INR / Each
Buy It Together
Current Item
Microscope Camera with Reduction Lens, C-Mount, 10 MP, USB 2.0
Microscope Camera with Reduction Lens, C-Mount, 10 MP, USB 2.0
7890514
₹93,100.00INR / Each
+ + + +
Microscope Camera Adapter for Nikon SLR/DSLR, 2X
7890500
₹32,800.00INR / Each
=
Total Price 1053900.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Includes
digital camera, 0.5X reduction lens, 6' (1.8 m) USB cable, 30 mm and 30.5 mm adapters, CD with software and user's instructions.
Key Features
  • Documentation and analysis for a deeper view of your slide
  • Easy to install and ready for immediate use
  • Capture, annotate, measure or evaluate still images and video
  • Digital camera features frame rate of 27 fps at 872 x 654, 8 fps at 1744 x 1308, and 1.9 fps at 3584 x 2748
More About this Item
AmScope™ microscope cameras are designed for ease of use and the ability to output images and video. These cameras can produce still image and video capture, still image measurement, and allow you to make notes, shapes, and watermarks with ease.
Cameras are available in a wide range of resolution and connectivity options for applications ranging from education, conservation, pharmaceutical, or other demanding fields. Microscopes sold separately.
Designed using the C-mount standard, the camera with the included reduction lens and two sizing-adapters can fit 23mm, 30mm, and 30.5mm photo ports and ocular ports. The reduction lens increases the field-of-view to better match what's seen through the eyepieces. Using a standard USB 2.0 connection, the camera is easy to install. Drawing power from the USB connection, no additional connections are needed.
1 Year Warranty
Specifications
  • Resolution10 MP, 3584 x 2748
  • Objectives0.5X reduction lens
  • Image Sensor1/2.3" Aptina MT9J001 color CMOS
  • OutputUSB 2.0
  • System RequirementsWindows XP/Vista/7/8/10 (32 & 64 bit), Mac OS X, Linux
  • Data TransferUSB 2.0
  • Power5 VDC over USB Port
  • DescriptionMicroscope Camera with Reduction Lens, C-Mount, 10 MP, USB 2.0