Microscope Camera with Reduction Lens and Calibration Slide, C-Mount, High-Speed, 5 MP, USB 2.0
Representative image only.
Microscope Camera with Reduction Lens and Calibration Slide, C-Mount, High-Speed, 5 MP, USB 2.0
| Item: 7890513
Capture images and stream video directly from your microscope
₹72,300.00INR / Each
₹72,300.00INR / Each
Buy It Together
Current Item
Microscope Camera with Reduction Lens and Calibration Slide, C-Mount, High-Speed, 5 MP, USB 2.0
Microscope Camera with Reduction Lens and Calibration Slide, C-Mount, High-Speed, 5 MP, USB 2.0
7890513
₹72,300.00INR / Each
+ + + +
Microscope Camera Adapter for Nikon SLR/DSLR, 2X
7890500
₹32,800.00INR / Each
=
Total Price 1033100.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Includes
digital camera, calibration slide, 0.5X reduction lens, 6' (1.8 m) USB cable, 30 mm and 30.5 mm adapters, CD with software and user's instructions.
Key Features
  • Documentation and analysis for a deeper view of your slide
  • Easy to install and ready for immediate use
  • Capture, annotate, measure or evaluate still images and video
  • High-speed digital camera features frame rate of 90 fps at 640 x 480, 27 fps at 1280 x 960, and 7 fps at 2592 x 1944
More About this Item
AmScope™ microscope cameras are designed for ease of use and the ability to output images and video. These cameras can produce still image and video capture, still image measurement, and allow you to make notes, shapes, and watermarks with ease.
Cameras are available in a wide range of resolution and connectivity options for applications ranging from education, conservation, pharmaceutical, or other demanding fields. Microscopes sold separately.
Designed using the C-mount standard, the camera with the included reduction lens and two sizing-adapters can fit 23mm, 30mm, and 30.5mm photo ports and ocular ports. The reduction lens increases the field-of-view to better match what's seen through the eyepieces. Using a standard USB 2.0 connection, the camera is easy to install. Drawing power from the USB connection, no additional connections are needed. The included calibration slide allows you to take accurate measurements in the software by creating baseline measurements at multiple magnifications.
1 Year Warranty
Specifications
  • Resolution5 MP, 2592x1944
  • Objectives0.5X reduction lens
  • Image Sensor1/2.5" Aptina MT9P001 color CMOS
  • OutputUSB 2.0
  • System RequirementsWindows XP/Vista/7/8/10 (32 & 64 bit), Mac OS X, Linux
  • Data TransferUSB 2.0
  • Power5 VDC over USB Port
  • DescriptionMicroscope Camera with Reduction Lens and Calibration Slide, C-Mount, High-Speed, 5 MP, USB 2.0