Mettler Toledo Versatile Pro pH Electrode; BNC with 3.9 ft Cable

Representative image only.

Mettler Toledo Versatile Pro pH Electrode; BNC with 3.9 ft Cable

Mettler Toledo | Mfr: 51343031| Item: 3590034

Expertly-engineered probes to cover a wide range of applications

₹130,000.00INR / Each

₹130,000.00INR / Each

Buy It Together

Current Item

Mettler Toledo Versatile Pro pH Electrode; BNC with 3.9 ft Cable
Mettler Toledo Versatile Pro pH Electrode; BNC with 3.9 ft Cable
3590034

₹130,000.00INR / Each
+ + + +
Cole-Parmer Adjustable-Volume Pipettor, 500 to 5000 µL
2450109

₹8,400.00INR / Each
=
Total Price 448700.00 ADD 5 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • ⌡ARGENTHAL™ reference system

More About this Item

These ISM® - Intelligent Sensor Management electrodes models store calibration data and automatically transfer the data to your meter. Choose open junction versions for dirty and complex samples. Cables not included with all electrodes. For additional versatility—select the one that connects to your meter.

ARGENTHAL™ reference system has a silver ion trap, for silver-ion-free electrolyte. No clogging junction from sulfides or protein binding—perfect for biological samples and TRIS buffers.

Specifications
  • ApplicationsGeneral-use
  • Electrode Body MaterialPolysulfone (PS)
  • Reference JunctionCeramic
  • Reference Electrolyte3M KCl
  • Sensing Glass TypeU - Universal for standard applications
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Temperature CompensationYes
  • GuardYes
  • ATC ConnectorRCA (cinch)
  • ATC ElementNTC 30 kΩ
  • ConnectorBNC
  • Diameter (mm)12
  • Length (mm)120
  • Cable Length (m)1.2
  • DescriptionpH Electrode; BNC with 3.9 ft Cable
Reviews
Q&A