Mettler Toledo STARe Software Silver Package
Representative image only.
Mettler Toledo STARe Software Silver Package
Mettler Toledo | Mfr: 30386626| Item: 2575354
Order accessories and software for more efficient DSC and TGA analysis
₹9,748,000.00INR / Each
₹9,748,000.00INR / Each
Buy It Together
Current Item
Mettler Toledo STARe Software Silver Package
Mettler Toledo STARe Software Silver Package
2575354
₹9,748,000.00INR / Each
+
Mettler Toledo High Temp DSC 3 with Sample Robot; 115 VAC, 60 Hz
2575340
₹30,254,000.00INR / Each
+ =
Total Price 58895000.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Great flexibility with unlimited evaluation possibilities
  • One database with five programs
  • Reliable automation to improve efficiency
  • Save time with easy OnceClick™ operation
  • 1 year warranty
More About this Item
STARe is a complete thermal analysis software with enormous flexibility and unlimited evaluation possibilities. Software utilizes 5 main programs linked to one another via the database: Installation, Module Control, Method, Experiment, and Evaluation Window.
1 Year Warranty
Specifications
  • DescriptionSTARe Software Silver Package