Mettler Toledo Thermogravimetry Analyzers
Characterize sample materials over a very wide temperature range
  • Ultra-micro balance provides exceptional accuracy, resolution, and repeatability
  • Built-in flow controller allows samples to be analyzed in defined atmospheres
  • OneClick™ offers fast and easy operation
Thermogravimeters measure the change in weight of a sample over time as it is heated, cooled, or held at constant temperature. This measurement provides valuable information of the sample such as phase transitions, adsorption, absorption, and thermal decomposition. Thermogravimeters are available with differential scanning calorimeters to analyze the thermal effects at the same time.
Easily view the terminal with the OneClick function plus the information on the status of the measurement even at a distance on the large display. Automatic buoyancy compensation eliminates the need to run a blank measurement allowing for faster accurate results. Use the built-in mass flow controllers to define the furnace atmosphere, which allows analysis of samples under a variety of atmospheres.
18,893,000.00 - ₹44,250,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Measurement Reproducibility Measurement Types Description
Availability Pricing
Mettler Toledo TGA2 Thermogravimetric Analyzer with Small Furnace, XP1 Balance, and TGA Sensor 2575344 Mfr # 30319250
COMPARE
<0.001 mg TGA sensor TGA2 Thermogravimetric Analyzer with Small Furnace, XP1 Balance, and TGA Sensor
₹18,893,000.00INR / Each
Mettler Toledo TGA/DSC 3+ with Large Furnace, XP1 Balance, and DTA Sensor 2575345 Mfr # 30319252
COMPARE
<0.001 mg DTA sensor TGA/DSC 3+ with Large Furnace, XP1 Balance, and DTA Sensor
₹25,922,000.00INR / Each
Mettler Toledo TGA/DSC 3+ with Large Furnace, XP1 Balance, Sample Robot, and SDTA Sensor 2575346 Mfr # 30421562
COMPARE
<0.001 mg SDTA sensor TGA/DSC 3+ with Large Furnace, XP1 Balance, Sample Robot, and SDTA Sensor
₹36,091,000.00INR / Each
Mettler Toledo TGA/DSC 3+ with Large Furnace, XP5 Balance, and SDTA Sensor 2575347 Mfr # 30319254
COMPARE
<0.002 mg SDTA sensor TGA/DSC 3+ with Large Furnace, XP5 Balance, and SDTA Sensor
₹29,029,000.00INR / Each
Mettler Toledo TGA/DSC 3+ with High-Temp Furnace, XP5U Balance, Sample Robot, and DSC Sensor 2575348 Mfr # 30293533
COMPARE
<0.0009 mg DSC sensor TGA/DSC 3+ with High-Temp Furnace, XP5U Balance, Sample Robot, and DSC Sensor
₹44,250,000.00INR / Each