Mettler Toledo Differential Scanning Calorimetry Analyzers

Measure enthalpy changes in samples with the highest accuracy and reproducibility
  • MultiSTAR™ sensor with 56 thermocouples helps detect the smallest and largest thermal effects
  • FlexCal™ calibration saves time and helps provide accurate measurements
  • OneClick™ feature offers fast and easy operation
Differential scanning calorimetry (DSC) is a thermal analysis technique that measures the amount of heat a sample absorbs or releases during heating or cooling. The amount of change in heat due to changes in physical and chemical properties of a sample is measured as a function of temperature or time. DSC instruments offer fast and easy way to detect transitions such as melts and phase changes, making them ideal for analytical, process control, quality assurance, and R&D laboratories.

Mettler Toledo DSC instruments feature innovative MultiSTAR sensor with 56 thermocouples for unbeatable sensitivity, outstanding resolution, and a perfectly flat baseline. The sensor with durable chemical-resistant ceramic surface offers low thermal mass and high conductivity resulting in excellent performance and long life span. The star-shaped arrangement of the thermocouples around the sample and reference crucibles guarantees flat baselines and reproducible results.

FlexiCal calibration automatically applies the correct adjustment parameters ensuring precise and accurate measurements. Store repetitive measurements and recall with a push of a key using the OneClick feature. Easily view the terminal with the OneClick function plus the information on the status of the measurement even at a distance on the large display. Select from models with sample robot to automate the process up to 34 samples at once. Select models with intracooler for applications requiring sample temperatures to –85°C.
22,565,000.00 - ₹35,464,000.00INR / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Mettler Toledo High Temp DSC 3 with Sample Robot; 115 VAC, 60 Hz 2575340 Mfr # 30421563
COMPARE
High Temp DSC 3 with Sample Robot; 115 VAC, 60 Hz
₹30,254,000.00INR / Each
Mettler Toledo High Temp DSC 3+ with Intracooler; 115 VAC, 60 Hz 2575341 Mfr # 30319177
COMPARE
High Temp DSC 3+ with Intracooler; 115 VAC, 60 Hz
₹22,565,000.00INR / Each
Mettler Toledo High Temp DSC 3+ with Sample Robot and Intracooler; 115 VAC, 60 Hz 2575343 Mfr # 30319178
COMPARE
High Temp DSC 3+ with Sample Robot and Intracooler; 115 VAC, 60 Hz
₹35,464,000.00INR / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat