Mettler Toledo STARe Software Option TGA Evaluation
Representative image only.
Mettler Toledo STARe Software Option TGA Evaluation
Mettler Toledo | Mfr: 119459| Item: 2575355
Order accessories and software for more efficient DSC and TGA analysis
₹2,276,000.00INR / Each
₹2,276,000.00INR / Each
Buy It Together
Current Item
Mettler Toledo STARe Software Option TGA Evaluation
Mettler Toledo STARe Software Option TGA Evaluation
2575355
₹2,276,000.00INR / Each
+
Mettler Toledo High Temp DSC 3 with Sample Robot; 115 VAC, 60 Hz
2575340
₹30,254,000.00INR / Each
+ =
Total Price 51423000.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Provides additional evaluation capabilities for thermogravimetric analysis
  • Features percent content, stoichiometric content, normalization to sample weight, conversion, and empirical content
  • 1 year warranty
1 Year Warranty
Specifications
  • DescriptionSTARe Software Option TGA Evaluation