Mettler Toledo Routine Pro-ISM pH Electrode; MultiPin™

Representative image only.

Mettler Toledo Routine Pro-ISM pH Electrode; MultiPin™

Mettler Toledo | Mfr: 51344055| Item: 3590022

Expertly-engineered probes to cover a wide range of applications

₹182,000.00INR / Each

₹182,000.00INR / Each

Buy It Together

Current Item

Mettler Toledo Routine Pro-ISM pH Electrode; MultiPin™
Mettler Toledo Routine Pro-ISM pH Electrode; MultiPin™
3590022

₹182,000.00INR / Each
+
Cole-Parmer Adjustable-Volume Pipettor, 500 to 5000 µL
2450109

₹8,400.00INR / Each
+ + + =
Total Price 500700.00 ADD 5 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • ISM® - Intelligent Sensor Management⌡Cable not included⌡ARGENTHAL™ reference system⌡HA glass membrane for high pH and temperature applications

More About this Item

These ISM® - Intelligent Sensor Management electrodes models store calibration data and automatically transfer the data to your meter. Choose open junction versions for dirty and complex samples. Cables not included with all electrodes. For additional versatility—select the one that connects to your meter.

ISM technology stores calibration data in the sensor and automatically transfer it to the meter.⌡Select the appropriate sensor cable for your application, meter, and ATC connection—BNC, waterproof BNC, DIN, and Lemo OO cables available with banana or RCA (cinch) connectors.⌡High alkali glass membrane provides low-alkali error—ideal for high pH and high temperature applications. ARGENTHAL™ reference system has a silver ion trap, for silver-ion-free electrolyte. No clogging junction from sulfides or protein binding—perfect for biological samples and TRIS buffers.

Specifications
  • ApplicationsHigh pH
  • Electrode Body MaterialGlass
  • Reference JunctionCeramic
  • Reference Electrolyte3M KCl
  • Sensing Glass TypeHA - High alkalifor high pH and high temp applications
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Temperature CompensationYes
  • ATC ElementNTC 30 kΩ
  • ConnectorMultipin™
  • Diameter (mm)12
  • Length (mm)120
  • DescriptionpH Electrode; MultiPin™
Reviews
Q&A