Mettler Toledo Micro Pro-ISM pH Electrode; MultiPin™

Representative image only.

Mettler Toledo Micro Pro-ISM pH Electrode; MultiPin™

Mettler Toledo | Mfr: 51344163| Item: 3590142

Expertly-engineered probes to cover a wide range of applications

₹290,000.00INR / Each

₹290,000.00INR / Each

Buy It Together

Current Item

Mettler Toledo Micro Pro-ISM pH Electrode; MultiPin™
Mettler Toledo Micro Pro-ISM pH Electrode; MultiPin™
3590142

₹290,000.00INR / Each
+ + + +
Cole-Parmer Adjustable-Volume Pipettor, 2000 to 10000 µL
2450110

₹8,400.00INR / Each
+
Mettler Toledo 51340024 Adapter, NS 14/15, 12 mm
3590395

₹6,000.00INR / Each
=
Total Price 619700.00 ADD 6 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • ISM® - Intelligent Sensor Management⌡Cable not included⌡ARGENTHAL™ reference system

More About this Item

These ISM® - Intelligent Sensor Management electrodes models store calibration data and automatically transfer the data to your meter. Choose open junction versions for dirty and complex samples. Cables not included with all electrodes. For additional versatility—select the one that connects to your meter.

ISM technology stores calibration data in the sensor and automatically transfer it to the meter.⌡Select the appropriate sensor cable for your application, meter, and ATC connection—BNC, waterproof BNC, DIN, and Lemo OO cables available with banana or RCA (cinch) connectors. ARGENTHAL™ reference system has a silver ion trap, for silver-ion-free electrolyte. No clogging junction from sulfides or protein binding—perfect for biological samples and TRIS buffers.

Specifications
  • ApplicationsSmall volumes and containers
  • Electrode Body MaterialGlass
  • Reference JunctionCeramic
  • Reference Electrolyte3M KCl
  • Sensing Glass TypeU - Universal for standard applications
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Temperature CompensationYes
  • ATC ElementNTC 30 kΩ
  • ConnectorMultipin™
  • Diameter (mm)5
  • Length (mm)130
  • DescriptionpH Electrode; MultiPin™
Reviews
Q&A