Mettler Toledo External Relay for Intracooler
Representative image only.
Mettler Toledo External Relay for Intracooler
Mettler Toledo | Mfr: 51143415| Item: 2575353
Order accessories and software for more efficient DSC and TGA analysis
₹364,000.00INR / Each
₹364,000.00INR / Each
Buy It Together
Current Item
Mettler Toledo External Relay for Intracooler
Mettler Toledo External Relay for Intracooler
2575353
₹364,000.00INR / Each
+
Mettler Toledo High Temp DSC 3 with Sample Robot; 115 VAC, 60 Hz
2575340
₹30,254,000.00INR / Each
+ =
Total Price 49511000.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Automate intracooler activity
  • 1 year warranty
More About this Item
Automatically sends signals to turn the intracooler on or off depending on the power state of the DSC.
1 Year Warranty
Specifications
  • Compatible WithMettler Toledo Intracooler 25753-52
  • DescriptionExternal Relay for Intracooler