Mettler Toledo D6 Excellence Density Meter, 100-240 VAC
Representative image only.
Mettler Toledo D6 Excellence Density Meter, 100-240 VAC
Mettler Toledo | Mfr: D6| Item: 1133530
Accurate, intuitive, and modular with the One Click user interface
₹13,139,000.00INR / Each
₹13,139,000.00INR / Each
Buy It Together
Current Item
Mettler Toledo D6 Excellence Density Meter, 100-240 VAC
Mettler Toledo D6 Excellence Density Meter, 100-240 VAC
1133530
₹13,139,000.00INR / Each
+
Mettler Toledo R4 Excellence Refractometer, 100-240 VAC
1133596
₹5,476,000.00INR / Each
=
Total Price 18615000.00 ADD 2 ITEMS TO CART
Product Description
Includes
Density meter, external power supply, country-specific power cable, terminal WVGA7 inch AnaChem, syringe adapter, waste tube, three disposable syringes, syringe holder Dx, combined water standard (9 mL), user manual, declaration of conformity, and test report.
Key Features
  • Powerful autosamplers for all sample types, including high-viscosity solutions
  • Simultaneous determination of density, refractive index, pH, conductivity and color
  • LabX PC software complies with 21 CFR Part 11
  • Accuracy of ±0.000005 g/cm3
  • 1 year warranty
More About this Item
Excellence Density Meters guarantee high reliability and maximum precision, with an accuracy of up to 6 digits. Measure density, specific gravity, as well as many related values such as % alcohol and Brix quickly. The touch screen terminal with intuitive One Click user interface minimizes training and operator influence. Ensure precise density measurements with the integrated video view and BubbleCheck™. Automatic viscosity correction combined with integrated temperature control ensures trustworthy results for all samples. The density meter and refractometer can be stacked, enabling you to combine a density meter with a refractive index cell to measure both parameters at once.
1 Year WarrantyConformite EuropeenneCanadian Standards Association
Specifications
  • Width (InchShort)15 5/32
  • Height (InchShort)15 5/32
  • Density Resolution (g/cm³)0.000001 g/cm3
  • Density Accuracy±0.000005 g/cm3
  • Width (cm)43.87
  • Width (in)15 5/32
  • DescriptionDensity meter Excellence D6