Mettler Toledo R4 Excellence Refractometer, 100-240 VAC
Representative image only.
Mettler Toledo R4 Excellence Refractometer, 100-240 VAC
Mettler Toledo | Mfr: R4| Item: 1133596
Excellence refractometers provide measurements for many challenging samples with an accuracy up to five decimal places
₹5,476,000.00INR / Each
₹5,476,000.00INR / Each
Product Description
Includes
Refractometer, external power supply, country-specific power cable, terminal WVGA 7-inch AnaChem, combined water standard (9 mL), three plastic pipettes, user manual, declaration of conformity, and test report.
Key Features
  • Measures in Brix and refractive index
  • Intuitive One Click™ user interface
  • Set up a range of acceptable limits on your samples for immediate QC results
  • Measurement range of 1.32 to 1.70 refractive index
  • 1 year warranty
More About this Item
These refractometers master measurements of a wide variety of challenging samples with high repeatability and an accuracy of up to five decimal places. The touch-screen terminal with the intuitive One Click™ user interface minimizes training and operator influence. Set up a range of acceptable limits for your samples; results that fall within this range are color-coded, enabling instant identification of accepted and rejected samples.
1 Year WarrantyConformite EuropeenneCanadian Standards Association
Specifications
  • Width (InchShort)15 5/32
  • Density Resolution (g/cm³)0.0001 nD
  • Height (InchShort)15 5/32
  • Density Accuracy±0.0001 nD
  • Width (cm)17.37
  • Width (in)15 5/32
  • DescriptionRefractometer Excellence R4