Mettler Toledo Excellence D-Series Density Meters
Accurate, intuitive, and modular with the One Click user interface
  • Powerful autosamplers for all sample types, including high-viscosity solutions
  • Simultaneous determination of density, refractive index, pH, conductivity and color
  • LabX PC software complies with 21 CFR Part 11
Excellence Density Meters guarantee high reliability and maximum precision, with an accuracy of up to 6 digits. Measure density, specific gravity, as well as many related values such as % alcohol and Brix quickly. The touch screen terminal with intuitive One Click user interface minimizes training and operator influence. Ensure precise density measurements with the integrated video view and BubbleCheck™. Automatic viscosity correction combined with integrated temperature control ensures trustworthy results for all samples. The density meter and refractometer can be stacked, enabling you to combine a density meter with a refractive index cell to measure both parameters at once.
9,596,000.00 - ₹13,139,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Mettler Toledo D5 Excellence Density Meter, 100-240 VAC 1133529 Mfr # 30474901
COMPARE
Density meter Excellence D5
₹9,596,000.00INR / Each
Mettler Toledo D6 Excellence Density Meter, 100-240 VAC 1133530 Mfr # D6
COMPARE
Density meter Excellence D6
₹13,139,000.00INR / Each