Mettler Toledo Crucible Sealing Press
Representative image only.
Mettler Toledo Crucible Sealing Press
Mettler Toledo | Mfr: 119410| Item: 2575357
Order accessories and software for more efficient DSC and TGA analysis
₹1,389,000.00INR / Each
₹1,389,000.00INR / Each
Buy It Together
Current Item
Mettler Toledo Crucible Sealing Press
Mettler Toledo Crucible Sealing Press
2575357
₹1,389,000.00INR / Each
+
Mettler Toledo High Temp DSC 3 with Sample Robot; 115 VAC, 60 Hz
2575340
₹30,254,000.00INR / Each
+ =
Total Price 50536000.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Use in DSC, TGA, and TMA techniques
  • Easily cold welds pan hermetically with the lid
  • Change the plunger and die to use with other crucibles
  • 1 year warranty
1 Year Warranty
Specifications
  • Diameter (InchShort)6
  • DescriptionCrucible Sealing Press