MadgeTech IFC300 USB Docking Station with USB Cable and Software.
Representative image only.
MadgeTech IFC300 USB Docking Station with USB Cable and Software.
MadgeTech | Mfr: IFC300| Item: 2300034
₹25,500.00INR / Each
₹25,500.00INR / Each
Buy It Together
Current Item
MadgeTech IFC300 USB Docking Station with USB cable and software.
MadgeTech IFC300 USB Docking Station with USB cable and software.
2300034
₹25,500.00INR / Each
+ + + =
Total Price 25500.00 ADD 4 ITEMS TO CART
Product Description
Key Features
  • Connect the data logger to PC for programming and data transfer
  • Required for operation of the logger
  • Designed for the Cryo-Temp, LyoTemp, and TransiTempII loggers
  • Features a single port
  • 1 year warranty
More About this Item
Enhance your data logging system with docking stations, mounting kits, thermal shields, and other accessories.
1 Year Warranty
Specifications
  • DescriptionUSB Docking Station with USB cable and software.