MadgeTech Cryo-Temp Ultra-Low Temperature Data Logger
Representative image only.
MadgeTech Cryo-Temp Ultra-Low Temperature Data Logger
MadgeTech | Mfr: CRYO-TEMP| Item: 2300027
TransiTempII, TransiTempII-RH, and Cryo-Temp
Call For Price
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Call For Price
Buy It Together
Current Item
MadgeTech Cryo-Temp Ultra-Low Temperature Data Logger
MadgeTech Cryo-Temp Ultra-Low Temperature Data Logger
2300027
Call For Price
+ + =
Total Price 70200.00 ADD 3 ITEMS TO CART
Product Description
Includes
Battery.
Key Features
  • Programmable alarms and warnings
  • Instantaneous out-of-range indication
  • IP65-rated splash proof housing
  • Three color LED status indicator
  • Delayed start
  • 1 year warranty
More About this Item
This splash-proof temperature data logger features a highly precise semiconductor temperature sensor and an ABS plastic enclosure. Use the data logger for monitoring and recording the temperature of all types of perishable products, including food items and pharmaceutical goods, both in storage and in transit. Order a USB docking station with USB cable and software 23000-34 to communicate with a computer. The unit features a handle for easy placement into a USB docking station.
The data logger handles temperatures down to 86°C and provides date and time stamped temperature readings. Easily view the data in graphical or tubular form. Set high and low alarm limits and high and low warning limits using the software. The three LED status indicators alert you to the temperature status: green signals safe conditions, yellow indicates warning limits have been exceeded, and red denotes alarm limits have been exceeded. The units can be programmed through software to start immediately, delayed start for up to 30 days, or be set to start using the magnetic switch.
For an NIST-traceable data logger, please contact the Customer Service Department.
IP 651 Year Warranty
Specifications
  • Number of Channels1
  • Input TypeTemperature
  • Min Temperature (° C)-86
  • Max Temperature (° C)35
  • Temperature Accuracy (° F)±2
  • Temperature Resolution (° C)0.1
  • Temperature Accuracy (° C)±1.0
  • Memory Size32,767
  • Battery Life1 year
  • Length (in)3 19/64
  • Height (in)2 7/64
  • Width (in)13/32
  • DescriptionUltra-Low Temperature Recorder