Lovibond MD 600 Multiparameter Colorimeter 214020

Representative image only.
Lovibond Mfr # 214020 – Item # WW-99599-00
Industry-leading 6 wavelength photodetector array allows for use of over 70 pre-programmed test methods
  • Internal memory stores up to 1000 readings—perfect for intensive field applications
  • Accepts Powder Pack™, tablets, liquid, and ampoule-based reagents for greater flexibility
  • Optional infrared data transfer module offers three interface options for added convenience

₹290,000.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Display Typegraphic display
  • Wavelength(s) (nm)430, 530, 560, 580, 610, 660, or automatic detection
  • Min Operating Temperature (° F)41
  • Max Operating Temperature (° F)104
  • Min Operating Temperature (° C)5
  • Max Operating Temperature (° C)40
  • OutputIR interface for data transfer, RJ45 connector for internet updates
  • Battery4 AA (included)
  • Length (in)8 1/4
  • Width (in)3 3/4
  • Height (in)1 5/8
  • Length (cm)21
  • Width (cm)9.4
  • Height (cm)4.4
  • DescriptionMultiparameter Colorimeter
Conformite Europeenne

Key Features

  • Internal memory stores up to 1000 readings—perfect for intensive field applications
  • Accepts Powder Pack™, tablets, liquid, and ampoule-based reagents for greater flexibility
  • Optional infrared data transfer module offers three interface options for added convenience

More About this Item

Versatile colorimeter tests for a wide range of water quality parameters via a 6-wavelength photodetector array. Over 70 pre-programmed test methods are present on the instrument and over 20 user-defined methods can be added for incredible measurement versatility. Meter automatically selects the wavelength for each test parameter. Use meter with a variety of test reagent platforms such as Powder Packs, tablets, liquid, or ampoule-based kits. Additional features include menu driven user interface with 7 language selection, large display, and rugged acid and solvent resistant keypad.

Expansive internal memory allows for storage of over 1000 readings. Data is easily transferred to a computer or printer via the optional IRiM Data Transfer Module (order separately). Data transfer module uses infrared technology to wirelessly beam readings from meter allowing for a fully waterproof system. Additional test methods can be downloaded to the data module via a computer and transferred to the meter for ultimate user configurability.

Refer to instruction manual for full specifications

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat