LaMotte SMART 3 1910 Multiwavelength Colorimeter, 100 to 240 V

Representative image only.

LaMotte SMART 3 1910 Multiwavelength Colorimeter, 100 to 240 V

LaMotte | Mfr: 1910| Item: 9956508

Select from over 80 preprogrammed tests or enter up to 10 user defined tests

Call For Price

Call For Price

Buy It Together

Current Item

LaMotte SMART 3  1910 Multiwavelength Colorimeter, 100 to 240 V
LaMotte SMART 3 1910 Multiwavelength Colorimeter, 100 to 240 V
9956508

Call For Price
+ +
Lovibond MD 600  Multiparameter Colorimeter 214020
9959900

₹533,000.00INR / Each
=
Total Price 533000.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

four 25-mm sample cells, USB cable and wall plug, and 1 rechargeable lithium-ion battery.

Key Features

  • Direct readout in concentration, percent transmittance, or absorbance—readings displayed simultaneously
  • cULus approved
  • 2 year warranty

More About this Item

Meter is easy to use. This colorimeter performs a variety of tests using the reagent test kits sold separately below. Logs up to 500 test results for easy recall.

Download data to a computer or printer via USB interface cable (included).

Two year warranty
Specifications
  • Length (")3 1/2
  • Display TypeLCD
  • Light SourceLED
  • Width (InchShort)2 1/2
  • Detector TypeSilicon Photodiode
  • Height (InchShort)7 1/2
  • Wavelength(s) (nm)425, 525, 568, or 635
  • OutputUSB
  • Battery1 rechargeable lithium-ion (included)
  • Power (VAC)100 to 240
  • Length (in)6
  • Width (in)3 1/4
  • Height (in)2 1/2
  • Length (cm)15.2
  • Width (cm)8.3
  • Height (cm)6.4
Reviews
Q&A