LaMotte DC-1500 Single Test Colorimeter Kit , Chlorine Free and Total " Liquid form

Representative image only.

LaMotte DC-1500 Single Test Colorimeter Kit , Chlorine Free and Total " Liquid form

LaMotte | Mfr: RBY 3240| Item: 9956212

Single test, direct-reading colorimeters feature microprocessor technology

Call For Price

Call For Price

Buy It Together

Current Item

LaMotte DC-1500 Single Test Colorimeter Kit , Chlorine Free and Total " Liquid form
LaMotte DC-1500 Single Test Colorimeter Kit , Chlorine Free and Total " Liquid form
9956212

Call For Price
+ +
Environmental Express Model 200 Chlorine Colorimeter
9956178

₹171,000.00INR / Each
+ =
Total Price 171000.00 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Includes

analyzer, glass sample vials, plastic water bottle, tablet crusher, test-specific reagent, foam-lined carrying case, and one 9 V battery.

Key Features

  • Analyzer features automatic wavelength selection for quick testing
  • 1 year warranty

More About this Item

Auto-zeroing and a hinged light cover make these meters easy to use. Ergonomic design fits in your hand comfortably. Water-resistant design protects your meter from damage. Download data to a computer or printer via the optional RS-232 interface.

1 Year Warranty
Specifications
  • Display TypeLCD
  • Light SourceLED
  • Detector TypeSilicon Photodiode
  • Test ParametersChlorine; free and total
  • Number of Tests Included144
  • Data LoggingYes
  • Battery1 9V (included)
  • Length (in)6 1/2
  • Width (in)3 1/4
  • Height (in)2 1/2
  • DescriptionSingle Test Colorimeter Kit , Chlorine Free and Total " Liquid form
  • Warranty1 year
Reviews
Q&A