Environmental Express Model 200 Chlorine Colorimeter
Representative image only.
Environmental Express Model 200 Chlorine Colorimeter
Environmental Express | Item: 9956178
Easily measure water samples for free and/or total chlorine in the field or on the bench
₹171,000.00INR / Each
₹171,000.00INR / Each
Buy It Together
Current Item
Environmental Express Model 200 Chlorine Colorimeter
Environmental Express Model 200 Chlorine Colorimeter
9956178
₹171,000.00INR / Each
+
HF Scientific 10478 Chlorine Pocket Colorimeter
9956130
₹181,000.00INR / Each
+ + + =
Total Price 580400.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Includes
100 free chlorine powder reagent packs, 100 total chlorine powder reagent packs, three glass (24 mm dia) sample cells with lid, two plastic (10 mm dia) sample cells with lid, light shielding rings, brush, carrying case, and four AAA batteries.
Key Features
  • Rugged, portable design adapts to use in the field or on your lab bench
  • Simplistic user interface for easy operation at any skill level
  • Preprogrammed testing ranges for free, total, and combined chlorine easily accessed for quick measurement (from 0.02 mg/L to 8 mg/L )
  • Single or multi point calibration to match simple or complex sample requirements
  • Auto zero function removes the need for re-zeroing the meter between sample measurements saving time in the field
  • IP68-rated waterproof design ensures watertight protection for up to 1 hour
  • High-quality optical system quickly delivers accurate reliable results removing the need for rechecks
  • Automatic storage of testing records for up to 16 readings with time-and-date stamp for accurate record keeping
  • Water proof IP 68 design
More About this Item
The Environmental Express CL 200 chlorine colorimeter is a single-parameter instrument for measuring the concentration of free and/or total chlorine in water samples. Its rugged, handheld design makes it adaptable for field use or in laboratory settings. With stored calibration ranges specific to chlorine measurement and a simplistic user interface, the meter is easily operated by technicians at any experience level, ensuring accurate results without the need for retesting.
Model CL 200 colorimeter is supplied ready to use with two preprogrammed testing ranges for free, total, and combined chlorine. Both ranges use the DPD testing method. When the instrument is turned on, it will automatically display the last test used with readings in ABS or %T units. Simply select the measuring range, prepare your sample, and test. Primary and secondary standards are available for on-site verification of instrument performance.
Specifications
  • Display TypeLCD
  • Light SourceLED
  • Wavelength(s) (nm)530 nm
  • Absorbance Accuracy3% full scale
  • Test ParametersChlorine
  • Min Operating Temperature (° F)41
  • Max Operating Temperature (° F)104
  • Min Operating Temperature (° C)5
  • Max Operating Temperature (° C)40
  • WaterproofIP68
  • Data LoggingAutomatic
  • Memory Size16 data sets with time and date stamp
  • Battery4 AA (included)
  • Range0 to 2.0 mg/L, 0 to 8.0 mg/L
  • DescriptionChlorine Colorimeter