Hach® DR300 Pocket Colorimeters
Single-parameter colorimeter you can take anywhere
  • Rugged design withstands water, drops, extreme temperatures, and dirt
  • Compact size fits easily into a kit or your pocket
  • Read the large digital display in any condition—features backlight
  • No need to calibrate—just blank and read!
  • Store and recall up to 50 measurements
The DR300 series continues Hach's legacy of single-parameter colorimeters for water testing. These units are factory calibrated, so you only need to zero the unit before taking readings. Secondary calibration sets are available for some parameters to meet regulatory requirements. New features include a more intuitive interface to reduce potential manual errors, and larger digits on the backlit display.
156,000.00INR / Each
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Hach DR 300 Chlorine Pocket Colorimeter; Free and Total, HR/LR WW-99573-75 Mfr # LPV445.97.00110
COMPARE
Bromine/Chlorine, Pocket Colorimeter; Free and Total
₹156,000.00INR / Each
Hach DR 300 Nitrate Pocket Colorimeter WW-99573-76 Mfr # LPV445.97.02110
COMPARE
Nitrate Pocket Colorimeter
₹156,000.00INR / Each
Hach DR 300 Dissolved Oxygen Pocket Colorimeter WW-99573-77 Mfr # LPV445.97.03110
COMPARE
Dissolved Oxygen Pocket Colorimeter
₹156,000.00INR / Each
Hach DR 300 Ozone Pocket Colorimeter WW-99573-78 Mfr # LPV445.97.04110
COMPARE
Ozone Pocket Colorimeter
₹156,000.00INR / Each
Hach DR 300 Phosphate Pocket Colorimeter WW-99573-79 Mfr # LPV445.97.06110
COMPARE
Phosphate Pocket Colorimeter
₹156,000.00INR / Each
Hach DR 300 Zinc Pocket Colorimeter WW-99573-80 Mfr # LPV445.97.09110
COMPARE
Zinc Pocket Colorimeter
₹156,000.00INR / Each
Hach DR 300 Molybdenum Pocket Colorimeter; LR/HR WW-99573-81 Mfr # LPV445.97.10110
COMPARE
Molybdenum Pocket Colorimeter; LR/HR
₹156,000.00INR / Each
Hach DR 300 Chlorine and pH Pocket Colorimeter WW-99573-82 Mfr # LPV445.97.12110
COMPARE
Chlorine and pH Pocket Colorimeter
₹156,000.00INR / Each
Hach DR 300 Manganese Pocket Colorimeter; HR WW-99573-83 Mfr # LPV445.97.15110
COMPARE
Manganese Pocket Colorimeter; HR
₹156,000.00INR / Each
Hach DR 300 Iron Pocket Colorimeter; TPTZ WW-99573-84 Mfr # LPV445.97.16110
COMPARE
Iron Pocket Colorimeter; TPTZ
₹156,000.00INR / Each
Hach DR 300 Iron Pocket Colorimeter; Ferrover WW-99573-85 Mfr # LPV445.97.22110
COMPARE
Iron Pocket Colorimeter; Ferrover
₹156,000.00INR / Each
Hach DR 300 Aluminum Pocket Colorimeter WW-99573-86 Mfr # LPV445.97.25110
COMPARE
Aluminum Pocket Colorimeter
₹156,000.00INR / Each
Hach DR 300 Monochloramine and Free Ammonia Pocket Colorimeter WW-99573-87 Mfr # LPV445.97.26110
COMPARE
Monochloramine and Free Ammonia Pocket Colorimeter
₹156,000.00INR / Each
Hach DR 300 Ammonia Nitrogen Pocket Colorimeter WW-99573-88 Mfr # LPV445.97.40110
COMPARE
Ammonia Nitrogen Pocket Colorimeter
₹156,000.00INR / Each
Hach DR 300 500 nm Pocket Colorimeter WW-99573-89 Mfr # LPV445.97.50110
COMPARE
500 nm Pocket Colorimeter
₹156,000.00INR / Each
Hach DR 300 Chlorine Dioxide Pocket Colorimeter WW-99573-90 Mfr # LPV445.97.51110
COMPARE
Chlorine Dioxide Pocket Colorimeter
₹156,000.00INR / Each
Hach DR 300 528 nm Pocket Colorimeter WW-99573-91 Mfr # LPV445.97.52110
COMPARE
528 nm Pocket Colorimeter
₹156,000.00INR / Each
Hach DR 300 600 nm Pocket Colorimeter WW-99573-92 Mfr # LPV445.97.60110
COMPARE
600 nm Pocket Colorimeter
₹156,000.00INR / Each
Hach DR 300 Chlorine Pocket Colorimeter; Free and Total, Mid-Range WW-99573-93 Mfr # LPV445.97.62110
COMPARE
Chlorine Pocket Colorimeter; Free and Total, Mid-Range
₹156,000.00INR / Each
Hach DR 300 655 nm Pocket Colorimeter WW-99573-94 Mfr # LPV445.97.65110
COMPARE
655 nm Pocket Colorimeter
₹156,000.00INR / Each
Showing 1- 20 of 20