Lovibond 215640 Verification Standard Kit for MD 600 Colorimeter
Representative image only.
Lovibond 215640 Verification Standard Kit for MD 600 Colorimeter
Lovibond | Mfr: 215640| Item: 9959904
Optimize water analysis
₹107,000.00INR / Each
₹107,000.00INR / Each
Product Description
Key Features
  • For Lovibond water analysis equipment
More About this Item
Choose the following water analysis accessories to meet your application needs.
The Verification Standard Kit for the MC500 is designed to confirm the accuracy and reliability of measurement results. Orbeco-Hellige recommends testing your instrument with this kit every three months to ensure the greatest accuracy.
Confirms the Standards of: Zero, 430, 530, 560, 580, 610, and 660 nm
Specifications
  • Compatible WithOrbeco-Hellige MC500 colorimeter
  • DescriptionVerification Standard Kit for MD 600 Colorimeter