Lovibond L147450 Test Kit , Alkalinity (Total), 20-240 mg/L
Representative image only.
Lovibond L147450 Test Kit , Alkalinity (Total), 20-240 mg/L
Lovibond | Mfr: 147450| Item: 0554230
Accurate and easy to use for routine water analysis
₹37,600.00INR / Each
₹37,600.00INR / Each
Buy It Together
Current Item
Lovibond L147450 Test Kit , Alkalinity (Total), 20-240 mg/L
Lovibond L147450 Test Kit , Alkalinity (Total), 20-240 mg/L
0554230
₹37,600.00INR / Each
+ =
Total Price 258600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
carrying case with instructions, comparator, color disc, sample cells, and reagents for 100 tests
Key Features
  • Continuous color disc
  • Tablet and powder reagent chemistries
  • 1 year warranty
More About this Item
Simply add the tablet or powder pack reagent to the sample cell, allow it to dissolve per specific parameter instructions, rotate the color disc to match the color of the sample, and read off the concentration value.
1 Year Warranty
Specifications
  • TestAlkalinity, total
  • Measurement Points20 to 240 ppm
  • Reagent PlatformTablet
  • Tests Per Kit100
  • DescriptionTest Kit , Alkalinity (Total), 20-240 mg/L