Lovibond Aqua Comparator Test Kits
Accurate and easy to use for routine water analysis
  • Continuous color disc
  • Tablet and powder reagent chemistries
Simply add the tablet or powder pack reagent to the sample cell, allow it to dissolve per specific parameter instructions, rotate the color disc to match the color of the sample, and read off the concentration value.
23,100.00 - ₹47,100.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Lovibond L147450 Test Kit , Alkalinity (Total), 20-240 mg/L WW-05542-30 Mfr # 147450
COMPARE
Test Kit , Alkalinity (Total), 20-240 mg/L
₹32,200.00INR / Each
Lovibond L147200 Test Kit , Aluminum, 0-0.3 mg/L WW-05542-31 Mfr # 147200
COMPARE
Test Kit , Aluminum, 0-0.3 mg/L
₹36,500.00INR / Each
Lovibond L147210 Test Kit , Ammonia, 0-1 mg/L WW-05542-32 Mfr # 147210
COMPARE
Test Kit , Ammonia, 0-1 mg/L
Call For Price
Lovibond L147052 Test Kit , Chlorine (Free and Total), 0-3.5 mg/L WW-05542-33 Mfr # 147052
COMPARE
Test Kit , Chlorine (Free and Total), 0-3.5 mg/L
₹26,400.00INR / Each
Lovibond L147051 Test Kit , Chlorine (Free), 0-3.5 mg/L WW-05542-34 Mfr # 147050
COMPARE
Test Kit , Chlorine (Free), 0-3.5 mg/L
₹23,100.00INR / Each
Lovibond L147050 Test Kit , Chlorine (Total), 0-3.5 mg/L WW-05542-35 Mfr # 147051
COMPARE
Test Kit , Chlorine (Total), 0-3.5 mg/L
₹25,000.00INR / Each
Lovibond L147490 Test Kit, Chlorine (Sodium Hypochlorite), 2 to 18% NaOCl WW-05542-36 Mfr # 147490
COMPARE
Test Kit , Chlorine (SOD ium Hypochlorite), 2-18% Naocl
₹47,100.00INR / Each
Lovibond L147330 Test Kit , Chlorine Dioxide, 0.01-0.2 mg/L WW-05542-38 Mfr # 147330
COMPARE
Test Kit , Chlorine Dioxide, 0.01-0.2 mg/L
₹27,000.00INR / Each
Lovibond L147430 Test Kit , Copper (Free and Total), 0-5 mg/L WW-05542-40 Mfr # 147430
COMPARE
Test Kit , Copper (Free and Total), 0-5 mg/L
₹32,200.00INR / Each
Lovibond L147320 Test Kit Iron, 1-10 mg/L WW-05542-42 Mfr # 147320
COMPARE
Test Kit Iron, 1-10 mg/L
₹27,000.00INR / Each
Lovibond L147290 Test Kit , Molybdate, 0-100 mg/L WW-05542-43 Mfr # 147290
COMPARE
Test Kit , Molybdate, 0-100 mg/L
₹37,400.00INR / Each
Lovibond L147480 Test Kit , Phosphate, 0-2.5 mg/L WW-05542-46 Mfr # 147480
COMPARE
Test Kit , Phosphate, 0-2.5 mg/L
Call For Price
Showing 1- 12 of 12