Lightnin 869440 Air-drive mixer with universal clamp

Representative image only.

Lightnin 869440 Air-drive mixer with universal clamp

Lightnin | Mfr: 869440| Item: 5032210

Use these mixers in the presence of volatile or explosive vapors, or where electrical power would be hazardous or impractical.

₹484,000.00INR / Each

₹484,000.00INR / Each

Buy It Together

Current Item

Lightnin 869440 Air-drive mixer with universal clamp
Lightnin 869440 Air-drive mixer with universal clamp
5032210

₹484,000.00INR / Each
+ =
Total Price 670000.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • 1 year warranty

More About this Item

These mixers are excellent for pilot studies of chemical, food, and wastewater treatment processes. We offer LABMASTER air-drive mixers with two types of mounting styles. The tube suspension mixers require the heavy-duty clamp 04560-24 (sold separately). The universal clamp mixers mount directly to a ring stand or tank rim.
Mixers include:
— 5/16" dia x 12"L, 316 SS nonthrough-type shaft
— 5/16" bore x 2.5" dia, 316 SS A-310 impeller
— collet-type chuck
— universal clamp or tube suspension mounting
— muffler
— needle value

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

1 Year Warranty
Specifications
  • Material316 Stainless Steel
  • Min Speed (rpm)30
  • Max Speed (rpm)1800
  • Mounting TypeC-clamp
  • Max Torque (in-oz)37
  • Motor TypeAir-driven
  • Max Air Inlet Pressure (PSI)90
  • Min Air Inlet Pressure (PSI)30
  • Shaft Diameter (in)0.75
  • Shaft Diameter (mm)19.1
  • Shaft Length (in)21.5
  • Shaft Length (mm)546
  • Motor hp1/15
  • DescriptionAir-drive mixer with universal clamp
  • Warranty1 Year
Reviews
Q&A